1-(5-Hydroxypyridin-3-yl)ethanone ≥95%

Supplier: Ambeed

A447487-100MG A447487-250MG
77616-934EA 152.44 USD
77616-934 77616-936
1-(5-Hydroxypyridin-3-yl)ethanone ≥95%
1-(5-Hydroxypyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.1360 g/mol
MDL Number: MFCD18256466
CAS Number: 910649-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR