5-Bromopyridine-2-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A439298-1G A439298-250MG A439298-100MG
77572-368EA 108.15 USD
77572-368 77594-376 77684-210
5-Bromopyridine-2-sulfonyl chloride ≥95%
5-Bromopyridine-2-sulfonyl chloride

Precaución: For research use only.

Formula: C₅H₃BrClNO₂S
MW: 256.51 g/mol
MDL Number: MFCD06739112
CAS Number: 874959-68-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR