GSK2830371 ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-5-(((5-Chloro-2-methylpyridin-3-yl)amino)methyl)-N-(3-cyclopentyl-1-(cyclopropylamino)-1-oxopropan-2-yl)thiophene-2-carboxamide, GSK 2830371

A447126-10MG A447126-1MG A447126-5MG
77616-928EA 75.44 USD
77616-928 77616-930 77616-932
GSK2830371 ≥98%
GSK2830371

Precaución: for Research Use Only

Formula: C₂₃H₂₉ClN₄O₂S
MDL Number: MFCD28009442
CAS Number: 1404456-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR