6-Fluoro-1H-indole-5-carbonitrile ≥97%

Supplier: Ambeed
Warning

A446476-1G A446476-100MG A446476-250MG A446476-5G
77613-374EA 436.72 USD
77613-374 77686-209 77686-210 77892-923
6-Fluoro-1H-indole-5-carbonitrile ≥97%

Precaución: for Research Use Only

Formula: C₉H₅FN₂
MDL Number: MFCD23706318
CAS Number: 1427358-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR