(S)-2-((tert-Butoxycarbonyl)amino)-3-mercaptopropanoic acid ≥95%

Supplier: Ambeed
Danger

Synonyms: (tert-Butoxycarbonyl)-D-cysteine

A438610-1G A438610-250MG
77613-344EA 95.79 USD
77613-344 77686-189
(S)-2-((tert-Butoxycarbonyl)amino)-3-mercaptopropanoic acid ≥95%
(S)-2-((tert-Butoxycarbonyl)amino)-3-mercaptopropanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₄S
MW: 221.27 g/mol
MDL Number: MFCD05663721
CAS Number: 149270-12-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR