5-Hydrazinyl-2-methoxypyridine dihydrochloride ≥97%

Supplier: Ambeed
Warning

A438526-1G A438526-5G
77594-366EA 23.58 USD
77594-366 77748-891
5-Hydrazinyl-2-methoxypyridine dihydrochloride ≥97%
5-Hydrazinyl-2-methoxypyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁Cl₂N₃O
MDL Number: MFCD26954811
CAS Number: 524699-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR