1-(5-Hydroxy-2-nitrophenyl)ethanone ≥95%

Supplier: Ambeed
Warning

A438379-1G A438379-250MG
77886-159EA 100 USD
77886-159 77886-160
1-(5-Hydroxy-2-nitrophenyl)ethanone ≥95%
1-(5-Hydroxy-2-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MW: 181.14 g/mol
MDL Number: MFCD12827423
CAS Number: 30879-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR