Rifamycin SV sodium ≥97%

Supplier: Ambeed
Warning

Synonyms: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione sodium salt, Rifamycin sodium

A445903-1G A445903-25G A445903-5G
77601-934EA 13.33 USD
77601-934 77601-936 77668-996
Rifamycin SV sodium ≥97%
Rifamycin SV sodium

Precaución: for Research Use Only

Formula: C₃₇H₄₆NNaO₁₂
MW: 719.8 g/mol
MDL Number: MFCD00056847
CAS Number: 14897-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR