1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥97%

Supplier: Ambeed
Danger

Synonyms: 1,2-Bis(phenylsulphinyl)ethanepalladium(II) acetate, 1,2-Bis(phenylsulphinyl)ethane palladium(II) acetate, White Catalyst, Christina White Catalyst

A445804-100MG A445804-250MG
77684-222EA 61 USD
77684-222 77684-223
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate ≥97%
1,2-Bis(phenylsulfinyl)ethanepalladium(II) diacetate

Precaución: For research use only.

Formula: C₁₆H₁₈O₄PdS₂
MW: 444.86 g/mol
MDL Number: MFCD09842752
CAS Number: 858971-43-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR