2-Bromo-6-fluoro-4-methylaniline ≥97%
Supplier: Ambeed
|
Warning
|
A437946-250MG
A437946-5G
A437946-1G
77616-884EA
11.33
USD
77616-884
77746-896
77746-895
2-Bromo-6-fluoro-4-methylaniline ≥97%
2-Bromo-6-fluoro-4-methylaniline
Precaución: for Research Use Only
Formula:
C₇H₇BrFN |
MDL Number:
MFCD07780634 CAS Number: 18349-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...