(1-Methylcyclobutyl)methanamine hydrochloride ≥98%

Supplier: Ambeed
Warning

A445447-10G A445447-1G A445447-250MG A445447-5G
77594-392EA 203.98 USD
77594-392 77594-394 77601-932 77872-724
(1-Methylcyclobutyl)methanamine hydrochloride ≥98%
(1-Methylcyclobutyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClN
MDL Number: MFCD18074449
CAS Number: 1245647-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR