(2-Formyl-5-(trifluoromethoxy)phenyl)boronic acid ≥97%

Supplier: Ambeed
Danger

Synonyms: 2-Formyl-5-(trifluoromethoxy)phenylboronic acid

A437239-1G A437239-250MG
77884-126EA 61 USD
77884-126 77884-127
(2-Formyl-5-(trifluoromethoxy)phenyl)boronic acid ≥97%
(2-Formyl-5-(trifluoromethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₆BF₃O₄
MW: 233.94 g/mol
MDL Number: MFCD10566600
CAS Number: 1218790-89-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR