Dimethyl difluoromalonate ≥95%

Supplier: Ambeed
Danger

A437023-5G A437023-25G
77601-888EA 9.23 USD
77601-888 77882-140
Dimethyl difluoromalonate ≥95%
Dimethyl difluoromalonate

Precaución: for Research Use Only

Formula: C₅H₆F₂O₄
MDL Number: MFCD00516931
CAS Number: 379-95-3
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR