Rifapentine ≥98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-3-(((4-Cyclopentyl-1-piperazinyl)imino)methyl)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione

A444634-100MG A444634-10MG A444634-250MG
77601-926EA 12.3 USD
77601-926 77601-928 77601-930
Rifapentine ≥98% (by HPLC)
Rifapentine

Precaución: for Research Use Only

Formula: C₄₇H₆₄N₄O₁₂
MW: 877.04 g/mol
MDL Number: MFCD00866806
CAS Number: 61379-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR