2-Cyano-6-methoxyphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Cyano-6-methoxyphenylboronic Acid

A444493-100MG A444493-1G A444493-250MG
77686-202EA 51 USD
77686-202 77686-203 77686-204
2-Cyano-6-methoxyphenylboronic acid ≥98%
2-Cyano-6-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₃
MW: 176.965 g/mol
MDL Number: MFCD10696677
CAS Number: 1164100-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR