7-(1-Acryloylpiperidin-4-yl)-2-(4-phenoxyphenyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxamide ≥99%

Supplier: Ambeed
Warning

Synonyms: (±)-Zanubrutinib

A444177-10MG A444177-1MG A444177-5MG
77616-902EA 113.3 USD
77616-902 77616-904 77616-906
7-(1-Acryloylpiperidin-4-yl)-2-(4-phenoxyphenyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxamide ≥99%
7-(1-Acryloylpiperidin-4-yl)-2-(4-phenoxyphenyl)-4,5,6,7-tetrahydropyrazolo[1,5-a]pyrimidine-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₇H₂₉N₅O₃
MW: 471.55 g/mol
MDL Number: MFCD30738015
CAS Number: 1633350-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR