Diallyl disulphide ≥80% stabilised

Supplier: Ambeed
Danger

Synonyms: Diallyldisulphide, Allyl disulphide

Stabilization: Stabilized with TBC

A444166-25G A444166-5G A444166-10G
77572-390EA 18 USD
77572-390 77601-922 77626-894
Diallyl disulphide ≥80% stabilised
Diallyl disulphide

Precaución: for Research Use Only

Formula: C₆H₁₀S₂
MDL Number: MFCD00008656
CAS Number: 2179-57-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR