1,7-Dimethyl-1H-indole-2,3-dione ≥95%

Supplier: Ambeed
Warning

A443775-1G A443775-250MG
77890-898EA 27 USD
77890-898 77890-899
1,7-Dimethyl-1H-indole-2,3-dione ≥95%

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MDL Number: MFCD00565658
CAS Number: 91790-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR