(S)-Benzyl 2,6-diaminohexanoate dihydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: H-Lys-OBzl·2HCl

A435129-1G A435129-100G
77673-062EA 10 USD
77673-062 77884-122
(S)-Benzyl 2,6-diaminohexanoate dihydrochloride ≥97%
(S)-Benzyl 2,6-diaminohexanoate dihydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₂Cl₂N₂O₂
MW: 309.23 g/mol
MDL Number: MFCD11501205
CAS Number: 16142-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR