Fmoc-L-β-homotryptophan ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-HoTrp-OH, (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(1H-indol-3-yl)butanoic acid

A435116-250MG A435116-1G A435116-100MG
77594-356EA 34.85 USD
77594-356 77601-880 77879-954
Fmoc-L-β-homotryptophan ≥95%
Fmoc-L-beta-homotryptophan

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₄
MDL Number: MFCD01863056
CAS Number: 353245-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR