(2R,3R)-3,5,7-Trihydroxy-2-(4-hydroxyphenyl)chroman-4-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Dihydrokaempferol

A435111-1MG A435111-5MG
77601-878EA 54.33 USD
77601-878 77668-942
(2R,3R)-3,5,7-Trihydroxy-2-(4-hydroxyphenyl)chroman-4-one ≥98%
(2R,3R)-3,5,7-Trihydroxy-2-(4-hydroxyphenyl)chroman-4-one

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₆
MW: 288.25 g/mol
MDL Number: MFCD00189454
CAS Number: 480-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR