2-Chloro-5-hydroxy-4-methylpyridine ≥97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-4-methylpyridin-3-ol, 2-Chloro-5-hydroxy-4-picoline

A443432-250MG A443432-5G
77613-362EA 33.99 USD
77613-362 77673-090
2-Chloro-5-hydroxy-4-methylpyridine ≥97%
2-Chloro-5-hydroxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.571 g/mol
MDL Number: MFCD16610471
CAS Number: 1227502-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR