cis-1,2-Cyclopentanedicarboxylic anhydride ≥97%

Supplier: Ambeed
Danger

Synonyms: cis-Tetrahydro-1H-cyclopenta[c]furan-1,3(3aH)-dione

A434627-1G A434627-5G A434627-250MG A434627-25G
77572-346EA 15 USD
77572-346 77594-352 77872-696 77872-697
cis-1,2-Cyclopentanedicarboxylic anhydride ≥97%
cis-1,2-Cyclopentanedicarboxylic anhydride

Precaución: for Research Use Only

Formula: C₇H₈O₃
MW: 140.14 g/mol
MDL Number: MFCD01311171
CAS Number: 35878-28-5
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR