5,7-Dihydroxy-2-(4-hydroxyphenyl)-6,8-bis((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Vicenin 2
A434186-1MG
A434186-10MG
A434186-5MG
77616-874EA
17.51
USD
77616-874
77748-879
77748-880
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6,8-bis((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one ≥97%
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6,8-bis((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one
Precaución: for Research Use Only
Formula:
C₂₇H₃₀O₁₅ MW: 594.52 g/mol |
MDL Number:
MFCD08458851 CAS Number: 23666-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...