Calcifediol monohydrate ≥98%

Supplier: Ambeed
Danger

Synonyms: (S,Z)-3-((E)-2-((1R,3aS,7aR)-1-((R)-6-Hydroxy-6-methylheptan-2-yl)-7a-methylhexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexanol hydrate, 25-Hydroxyvitamin D3

A443037-100MG A443037-1MG A443037-50MG
77686-198EA 262 USD
77686-198 77686-199 77686-200
Calcifediol monohydrate ≥98%
Calcifediol monohydrate

Precaución: for Research Use Only

Formula: C₂₇H₄₄O₂·H₂O
MW: 400.65 g/mol
MDL Number: MFCD08600001
CAS Number: 63283-36-3
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR