2-(2-Naphthoxy)ethanol ≥98%

Supplier: Ambeed
Warning

A433396-10G A433396-1G A433396-25G A433396-5G A433396-100G
77668-934EA 57 USD
77668-934 77668-936 77668-938 77668-940 77879-953
2-(2-Naphthoxy)ethanol ≥98%
2-(2-Naphthoxy)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₂
MDL Number: MFCD00016809
CAS Number: 93-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR