Rhodamine 123 ≥95%

Supplier: Ambeed
Warning

Synonyms: 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl xanthylium chloride, 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl)xanthylium chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester

A442351-50MG A442351-100MG A442351-1G A442351-250MG A442351-5G
77572-382EA 98 USD
77572-382 77601-916 77668-976 77668-978 77872-716
Rhodamine 123 ≥95%
Rhodamine 123

Precaución: for Research Use Only

Formula: C₂₁H₁₇ClN₂O₃
MDL Number: MFCD00012664
CAS Number: 62669-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR