3-Methoxycyclobutane-1-carboxylic acid ≥98%

Supplier: Ambeed
Danger

A442347-1G A442347-250MG
77884-141EA 281 USD
77884-141 77884-142
3-Methoxycyclobutane-1-carboxylic acid ≥98%
3-Methoxycyclobutane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MDL Number: MFCD08705860
CAS Number: 480450-03-1
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR