2-Bromo-6-iodonaphthalene ≥98%

Supplier: Ambeed
Danger

A433237-1G A433237-250MG
77886-148EA 91 USD
77886-148 77886-149
2-Bromo-6-iodonaphthalene ≥98%
2-Bromo-6-iodonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆BrI
MDL Number: MFCD17012551
CAS Number: 389806-32-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR