1-Phenyl-1H-imidazole-4-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A432960-1G A432960-250MG
77872-693EA 100 USD
77872-693 77872-694
1-Phenyl-1H-imidazole-4-carboxylic acid ≥97%
1-Phenyl-1H-imidazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD13193504
CAS Number: 18075-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR