Methyl 5-cyano-2-fluorobenzoate ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Cyano-2-fluorobenzoic acid methyl ester

A432905-5G A432905-1G
77572-342EA 82.4 USD
77572-342 77594-344
Methyl 5-cyano-2-fluorobenzoate ≥98%
Methyl 5-cyano-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₆FNO₂
MW: 179.148 g/mol
MDL Number: MFCD11042790
CAS Number: 337362-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR