2-Amino-4,5-dichlorophenol ≥97%

Supplier: Ambeed
Warning

A432833-100MG A432833-5G
77884-115EA 19 USD
77884-115 77884-116
2-Amino-4,5-dichlorophenol ≥97%
2-Amino-4,5-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MDL Number: MFCD12406838
CAS Number: 28443-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR