Triammonium citrate ≥99%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

A423625-100G A423625-25G
77613-310EA 6.18 USD
77613-310 77686-167
Triammonium citrate ≥99%
Triammonium citrate

Precaución: for Research Use Only

Formula: C₆H₁₇N₃O₇
MW: 243.22 g/mol
MDL Number: MFCD00036406
CAS Number: 3458-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR