Coumarin-6-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A432130-1G A432130-250MG
77594-340EA 91.23 USD
77594-340 77616-864
Coumarin-6-sulfonyl chloride ≥95%
Coumarin-6-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₅ClO₄S
MDL Number: MFCD01941320
CAS Number: 10543-42-7
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR