1H,1H,2H,2H-Heptadecafluorodecyl methacrylate ≥98% stabilised

Supplier: Ambeed
Warning

Synonyms: 2-(Perfluorooctyl)ethyl methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A431953-10G A431953-1G A431953-5G A431953-25G A431953-100G
77240-882EA 9 USD
77240-882 77240-884 77240-886 77746-874 77746-873
1H,1H,2H,2H-Heptadecafluorodecyl methacrylate ≥98% stabilised
1H,1H,2H,2H-Heptadecafluorodecyl methacrylate

Precaución: for Research Use Only

Formula: C₁₄H₉F₁₇O₂
MW: 532.2 g/mol
MDL Number: MFCD00042307
CAS Number: 1996-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR