Fmoc-(S)-3-amino-4-(3-cyanophenyl)butyric acid ≥98%

Supplier: Ambeed
Warning

A423340-1G A423340-100MG
77882-110EA 400 USD
77882-110 77882-109
Fmoc-(S)-3-amino-4-(3-cyanophenyl)butyric acid ≥98%
Fmoc-(S)-3-amino-4-(3-cyanophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₂₆H₂₂N₂O₄
MDL Number: MFCD01861077
CAS Number: 270065-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR