Ethyl-1-benzyl-4-piperidinecarboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylisonipecotic Acid Ethyl Ester, Ethyl 1-Benzyl-4-piperidinecarboxylate, 1-Benzyl-4-piperidinecarboxylic Acid Ethyl Ester, Ethyl 1-Benzylisonipecotate, Ethyl N-benzylpiperidine-4-carboxylate

A431864-100G A431864-25G A431864-10G A431864-500G A431864-5G
77601-866EA 59.45 USD
77601-866 77601-868 77668-920 77668-922 77668-924
Ethyl-1-benzyl-4-piperidinecarboxylate ≥98%
Ethyl-1-benzyl-4-piperidinecarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₂
MDL Number: MFCD00040748
CAS Number: 24228-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR