2-((1,3-Dimethyl-3,4,5,6-tetrahydropyrimidin-1-ium-2-yl)thio)pyridine 1-oxide hexafluorophosphate(V) ≥97%

Supplier: Ambeed
Warning

A431605-1G A431605-25G A431605-5G
77882-125EA 68 USD
77882-125 77882-126 77882-127
2-((1,3-Dimethyl-3,4,5,6-tetrahydropyrimidin-1-ium-2-yl)thio)pyridine 1-oxide hexafluorophosphate(V) ≥97%
2-((1,3-Dimethyl-3,4,5,6-tetrahydropyrimidin-1-ium-2-yl)thio)pyridine 1-oxide hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₁₁H₁₆F₆N₃OPS
MDL Number: MFCD08274620
CAS Number: 366821-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR