2-Hydroxyquinoline-6-sulfonyl chloride ≥95%

Supplier: Ambeed
Danger

A431310-1G A431310-250MG A431310-100MG
77886-144EA 260 USD
77886-144 77886-145 77886-143
2-Hydroxyquinoline-6-sulfonyl chloride ≥95%
2-Hydroxyquinoline-6-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₆ClNO₃S
MDL Number: MFCD18300284
CAS Number: 569340-07-4
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR