7-Methoxyindole-2-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 7-Methoxy-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A431159-1G A431159-5G A431159-100MG A431159-250MG
77613-330EA 287.37 USD
77613-330 77613-332 77884-111 77884-112
7-Methoxyindole-2-boronic acid pinacol ester ≥98%
7-Methoxyindole-2-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₀BNO₃
MW: 273.14 g/mol
MDL Number: MFCD11858357
CAS Number: 1072812-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR