1,16-Dibromohexadecane ≥98%

Supplier: Ambeed
Warning

A422716-1G A422716-5G A422716-25G
77616-832EA 65.92 USD
77616-832 77616-834 77746-851
1,16-Dibromohexadecane ≥98%
1,16-Dibromohexadecane

Precaución: for Research Use Only

Formula: C₁₆H₃₂Br₂
MDL Number: MFCD00039195
CAS Number: 45223-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR