Cobicistat ≥98%

Supplier: Ambeed
Warning

Synonyms: Thiazol-5-ylmethyl ((2R,5R)-5-((S)-2-(3-((2-isopropylthiazol-4-yl)methyl)-3-methylureido)-4-morpholinobutanamido)-1,6-diphenylhexan-2-yl)carbamate

A421342-5MG A421342-10MG A421342-1MG
77613-302EA 154.5 USD
77613-302 77613-298 77613-300
Cobicistat ≥98%
Cobicistat

Precaución: for Research Use Only

Formula: C₄₀H₅₃N₇O₅S₂
MW: 776.02 g/mol
MDL Number: MFCD18251449
CAS Number: 1004316-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR