6-Chloronaphthalen-1-ol ≥95%

Supplier: Ambeed
Warning

A430248-250MG A430248-100MG
77613-326EA 177.16 USD
77613-326 77890-880
6-Chloronaphthalen-1-ol ≥95%
6-Chloronaphthalen-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₇ClO
MDL Number: MFCD00466949
CAS Number: 56820-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR