Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥98%
Supplier: Ambeed
|
Warning
|
A430071-250MG
A430071-1G
77616-854EA
54.59
USD
77616-854
77746-870
Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) ≥98%
Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl][(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II)
Precaución: for Research Use Only
Formula:
C₅₈H₄₈Cl₂N₂P₂Ru MW: 1006.9387 g/mol |
MDL Number:
MFCD02684566 CAS Number: 212210-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...