1-Boc-3-(aminomethyl)-3-hydroxypyrrolidine ≥95%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 3-(aminomethyl)-3-hydroxypyrrolidine-1-carboxylate

A421008-1G A421008-250MG
77890-864EA 134 USD
77890-864 77890-865
1-Boc-3-(aminomethyl)-3-hydroxypyrrolidine ≥95%
1-Boc-3-(aminomethyl)-3-hydroxypyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.28 g/mol
MDL Number: MFCD18917057
CAS Number: 114214-73-2
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR