4-Bromo-7-nitro-1H-indazole ≥98%

Supplier: Ambeed
Danger

A430043-100MG A430043-250MG
77882-119EA 36 USD
77882-119 77882-120
4-Bromo-7-nitro-1H-indazole ≥98%
4-Bromo-7-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD12963530
CAS Number: 1190319-86-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR