(E)-(2-Methylstyryl)boronic acid ≥97%

Supplier: Ambeed
Warning

A430017-100MG A430017-250MG
77783-411EA 215 USD
77783-411 77783-412
(E)-(2-Methylstyryl)boronic acid ≥97%
(E)-(2-Methylstyryl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₂
MW: 161.99 g/mol
CAS Number: 1268631-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR