(2-Aminopyridin-3-yl)boronic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A430012-100MG A430012-1G A430012-250MG A430012-5G
77684-199EA 61 USD
77684-199 77886-137 77886-138 77886-139
(2-Aminopyridin-3-yl)boronic acid ≥98%, Ambeed.Inc
(2-Aminopyridin-3-yl)boronic acid

Precaución: For research use only.

Formula: C₅H₇BN₂O₂
MDL Number: MFCD11865292
CAS Number: 1204112-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR