(11aS)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-9-phenanthrenyl-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin ≥98%

Supplier: Ambeed
Warning

A420589-50MG A420589-100MG A420589-250MG A420589-1G
77746-849EA 125 USD
77746-849 77746-846 77746-848 77746-847
(11aS)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-9-phenanthrenyl-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin ≥98%
(11aS)-10,11,12,13-Tetrahydro-5-hydroxy-3,7-di-9-phenanthrenyl-5-oxide-diindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocin

Precaución: for Research Use Only

Formula: C₄₅H₃₁O₄P
MW: 666.7 g/mol
MDL Number: MFCD29905022
CAS Number: 1585988-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR