(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dimethylphenyl)propanoic acid ≥98%

Supplier: Ambeed
Warning

A420442-1G A420442-5G
77886-111EA 85 USD
77886-111 77886-112
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dimethylphenyl)propanoic acid ≥98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,4-dimethylphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₄
MDL Number: MFCD06659141
CAS Number: 1217728-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR